Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin, 9048-46-8, MFCD00130384, 5g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A88065G
Encompass
Fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Catalog No. 50-165-7297