Learn More
Selleck Chemical LLC Bovine Serum Albumin P1245-1G
SDP
Supplier:  Selleck Chemical LLC P12451G
Encompass_Preferred
Bovine serum albumin (BSA Albumin bovine serum) is an efficient passivating agent that inhibits nonspecific protein binding in assays such as ELISA It is used to coat hydrophobic substrates enhancing anti-fouling properties and reducing immune system interactions
Catalog No. 50-000-40071