Learn More
Bioworld Bovine Serum Albumin, Fraction V - Fine Powder 100g
Supplier:  Bioworld 220700162
Encompass_Preferred
Standard grade BSA is a highly purified, lyophilized powder. Its characteristics include minimum 98% albumin, pH adjusted to 7.0, caprylate-free, and low conductivity. It is used in bacterial media, enzyme systems, and as a protein base. Appearance Form : Powder, Appearance Color : White to light brown. 100g
Catalog No. 50-253-917