Learn More
Bioworld Bovine Serum Albumin (BSA), Fraction V - Low Endotoxin, 100 g
Supplier:  Bioworld 220700192
Encompass_Preferred
Bovine Serum Albumin (BSA), Fraction VIdeal for most cell culture applications and assays that require low endotoxin levelsThis product is processed to provide a minimum of 98% albumin and to be essentially free of protease, IgG and non-esterified fatty acids Endotoxin content is typically less than 0.5 EU/mgSpecificationspH: 7.0Low endotoxinhighly purified lyophilized powderShips at room temperature
Catalog No. 50-253-923