Learn More
Gold Biotechnology Inc Bovine Serum Albumin (BSA), Fraction V, Protease Free 1 kg
SDP
Supplier:  Gold Biotechnology Inc A4201
Encompass_Preferred
GoldBio BSA is a highly purified, lyophilized albumin, suitable for Westerns, enzyme systems, protein standards, hybridizations and as a protease-sensitive immunoassays. It is also perfectly suitable as a nutrient in cell culture media, with low interference and backgrounds and is ideal for most standard molecular biology experiments. BSA protein is a single polypeptide chain consisting of ~583 amino acid residues and no carbohydrates and is approximately 66 kDa in size. Fraction V refers to the fifth fraction of the original Cohn purification of plasma proteins using cold ethanol precipitation and is still commonly used as a name for bovine serum albumin regardless of purification method.
Catalog No. NC1391859