Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin chrom100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A1933100G
Encompass
Bovine serum albumin (BSA) is a 66 kDa globular protein crucial for transporting molecules like fatty acids drugs and hormones from the bloodstream. BSA is a drug carrier for bioactive compounds and has wide clinical applications. BSA has extensive usage in biological research including cell culture molecular biology protein biochemistry and detection techniques. It has wide pharmaceutical applications due to ease of production and interaction flexibility.
Catalog No. 50-290-8359