Learn More
Bioworld Bovine Serum Albumin (BSA), Heat Shock, Standard, 50 g
Supplier:  Bioworld 220700411
Encompass_Preferred
Prepared by a proprietary heat-shock process.An economical powder for a wide range of applications including use as a blocking agent,diluent,ELISAs,and immunoblotsPurity:>96%.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-253-964