Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin heat1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A96471KG
Encompass
Bovine serum albumin is a protein whose key biological function is to regulate the colloidal osmotic pressure of blood. BSA is widely used in various studies and applications. Made up of 583 amino acids BSA is a water-soluble protein with a calculated molecular weight of 66 430 Da. Six helices form three homologous domains of BSA. Depending on pH it undergoes reversible conformational isomerization. The native structure of the protein becomes reactive and flexible on heating.
Catalog No. 50-290-8167