Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin lyoph100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A3608100G
Encompass
Bovine serum albumin (BSA) is a 66 kDa protein consisting of three domains with two subdomains under each. It is a helical globular and non-glycosylated protein. It belongs to the serum albumins family. It has 17-disulfide bonds. Albumin is the most abundant plasma protein in humans.
Catalog No. 50-290-3047