Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin lyoph25G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A461225G
Encompass
The most abundant plasma protein in mammals is albumin. It is produced in the liver and transferred as a non-glycosylated protein into the plasma. This multifunctional protein has an amazing ability to bind to ligands.
Catalog No. 50-290-8221