Learn More
ALADDIN SCIENTIFIC CORPORATION Bovine Serum AlbuminBSA, 5g
Supplier:  ALADDIN SCIENTIFIC CORPORATION B2659935G
Encompass_Preferred
New Zealand Precision Grade Bovine Serum Albumin product is an ideal choice for diagnostic and research applications where enhanced sensitivity is required due to low NEFA and high Monomer specifications ApplicationsDiagnostic amp research applications where enhanced sensitivity is required due to low NEFA specificationIdeal for sensitive IVD assays susceptible to background interferenceProtease-sensitive immunoassays such as RIA EIA fluorescent andchemiluminescentProtein standard diluentProtein conjugate or enzyme stabilizerHybridizationCell culture applications Including development of serum-free mediaFeatures/BenefitsHigh purity low interference low backgroundFatty acid freeVirtually no detectable protease activity ensures Assay Integrity Closed Loop manufacturing minimizes open air contamination amp maximizes lot-to-lot consistencySuperior solubility/filterability for ease of useManufactured according to FDA cGMPsSpecificationsProtein (dry basis) 98 0percentPurity (albumin) 98p
Catalog No. 50-000-82760