Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine srum Albumin solut250ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A3299250ML
Encompass
Bovine serum albumin (BSA)nbspis a member of the serum albuminnbspfamily. It is anbspglobular helical and non-glycosylatednbspprotein. BSA is a single-chain protein and consists ofnbspthree domains with two sub-domains under eachItnbsphas 17-disulfidenbspbonds.
Catalog No. 50-290-3109