Learn More
American Research Products Inc BoVine transforming growth fac
Supplier:  American Research Products Inc CSBE14208B
Encompass
BoVine transforming growth factor b1 TGF-b1 ELISA Kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-29578