Learn More
Medchemexpress LLC Bovinic acid | 2540-56-9 | 96.2% | 280.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY113162
Encompass_Preferred
Bovinic acid is an orally active anti-inflammatory agent that inhibits oxidative stress and ferroptosis by regulating the Keap1-Nrf2 signaling pathway. It also has hepatoprotective effects against alcohol-associated liver disease.
  • Inhibits oxidative stress and ferroptosis in HepG2 cells.
  • Activates the Keap1-Nrf2 antioxidant signaling pathway.
  • Alleviates ethanol-induced alcohol-related liver disease.
  • Reduces elevated serum ALT and AST levels.
  • Increases hepatic superoxide dismutase (SOD) activity.
  • Decreases hepatic malondialdehyde (MDA) levels.
  • Reduces hepatic Fe2+ levels.
  • Improves liver parenchymal morphology.
Catalog No. 50-003-64342