Learn More
Andwin Scientific BOWIE-DICK + PEST 6/BOX
SDP
Supplier:  Andwin Scientific 34010105
Encompass_Preferred
The 3M Comply Bowie-Dick Plus Test Packs can be utilized to facilitate the performance of vacuum-assisted 132 to 134�C (270 to 273�F) steam sterilizers. This product is able to detect residual air in the sterilizer chamber due to air leaks, air removal failures, and inadequate vacuum. The test sheets are easily readable and can be kept for filing. This is sold in a pack of 6.
Catalog No. NC0492483