Learn More
Medchemexpress LLC Bowman-birk inhibitor | 37330-34-0 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP302610MG
Encompass_Preferred
The Bowman-Birk inhibitor is a highly cross-linked protein with seven disulfide bridges. It has distinct domains designed to inhibit both trypsin and chymotrypsin, serving as a significant plant protease inhibitor with anticarcinogenic properties.
  • Highly cross-linked protein with seven disulfide bridges
  • Inhibits both trypsin and chymotrypsin
  • Acts as a plant protease inhibitor
  • Possesses anticarcinogenic properties
  • Targets Ser/Thr protease and endogenous metabolite pathways
Catalog No. 50-003-96595