Learn More
Strem, An Ascensus Company CAS# 1656253-81-4. 25mg. (4R,4'R,5S,5'S)-2,2'-(1,3-Dihydro-2H-inden-2-ylidene)bis[4,5-dihydro-4,5-diphenyloxazole], 98%.
SDP
Supplier:  Strem, An Ascensus Company 07108525MG
Encompass
CAS# 1656253-81-4. 25mg. (4R,4'R,5S,5'S)-2,2'-(1,3-Dihydro-2H-inden-2-ylidene)bis[4,5-dihydro-4,5-diphenyloxazole], 98%. . Molecular Weight: 560.7. Molecular Formula: C39H32N2O2. Color/form: off white to light yellow pwdr. Strem# 07-1085. http://www.strem.com/catalog/v/07-1085/
Catalog No. 50-206-5400