Learn More
Thomas Scientific Freezer Box, 100 X 1.5 ml, Orange
SDP
Supplier:  Thomas Scientific 2591H29
Encompass
100 place box with hinged lid Polypropylene construction Will not degrade or absorb moisture Withstand freezing (to -80°C) and autoclaving Boxes hold 100 1.5/2.0 mL microtubes. The translucent lid is alphanumerically labeled for easy sample identification. A three-point hinge maintains its integrity during repeated use and a positive catch keeps lid securely fastened. Dimensions: 142 x 142 x 55 mm.
Catalog No. 50-146-1064