Learn More
Selleck Chemical LLC Box5-5mg
SDP
Supplier:  Selleck Chemical LLC P12165MG
Encompass_Preferred
Box5, a Wnt5a-derived N-butyloxycarbonyl hexapeptide (t-Boc-Met-Asp-Gly-Cys-Glu-Leu), acts as an Wnt5a antagonist, inhibits the basal migration and invasion of Wnt5a-expressing HTB63 melanoma cells, antagonizes the effects of Wnt5a on melanoma cell migration and invasion by directly inhibiting Wnt5a-induced PKC and Ca(2+) signaling.
Catalog No. 50-238-9837