Learn More
Medchemexpress LLC Bozitinib | 1440964-89-5 | 98.9% | 424.38 | C20H15F3N8 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12501750MG
Encompass_Preferred
Bozitinib is a selective, ATP-competitive c-MET (HGFR) kinase inhibitor with reported blood-brain barrier permeability, supplied for preclinical research applications. It is provided with defined chemical characterization and high purity suitable for in vitro and in vivo studies.
  • Selective c-MET kinase inhibition.
  • Demonstrated blood-brain barrier permeability.
  • Suitable for in vitro and in vivo preclinical research.
  • High chemical purity (98.9%).
  • Molecular formula C20H15F3N8; molecular weight 424.38 g/mol.
  • Available in multiple pack sizes including 50 mg.
Catalog No. 50-002-37431