Learn More
Medchemexpress LLC HY-125017 5mg Medchemexpress, Bozitinib CAS:1440964-89-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1250175MG
Encompass_Preferred
Medchemexpress, HY-125017 5mg Bozitinib CAS:1440964-89-5 Bozitinib (PLB-1001) is a highly selective c-MET kinase inhibitor with blood-brain barrier permeability. Bozitinib (PLB-1001) is a ATP-competitive small-molecule inhibitor, binds to the conventional ATP-binding pocket of the tyrosine kinase superfamily. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9685