Learn More
Selleck Chemical LLC BP-1-102 5mg 1334493-07-0
SDP
Supplier:  Selleck Chemical LLC S77695MG
Encompass_Preferred
BP-1-102 is a potent, orally bioavailable and selective STAT3 inhibitor, binds Stat3 with an affinity Kd of 504 nM and blocks Stat3-phospho-tyrosine (pTyr) peptide interactions and Stat3 activation at 4-6.8 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5225