Learn More
Medchemexpress LLC BP 897 hydrochloride 10mM 1mL | 314776-92-6 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY10666010MM 1ML
Encompass_Preferred
BP 897 hydrochloride is a potent and partial dopamine D3 receptor agonist and a weak D2 receptor antagonist BP 897 hydrochloride displays a high affinity at the dopamine D3 receptor (Ki 0 92 nM) and a 70 times lower affinity at the D2 receptor (Ki 61 nM)[1
Catalog No. 50-002-40728