Learn More
Medchemexpress LLC BPH-652 | 157124-84-0 | 99.0% | 500.63 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY16115
Encompass_Preferred
BPH-652 is a *S. aureus* dehydrosqualene synthase (CrtM) inhibitor. It has a Ki of 1.5 nM and an IC50 of 100-300 nM (*S. aureus* pigment formation). BPH-652 treatment has been shown to significantly lower *S. aureus* bacterial counts in the kidneys of mice in a systemic *S. aureus* infection model.
  • Inhibitor of *S. aureus* dehydrosqualene synthase (CrtM)
  • Ki of 1.5 nM (CrtM)
  • IC50 of 100-300 nM (*S. aureus* pigment formation)
  • Significantly lowers *S. aureus* bacterial counts in infected mice
  • For research use only
Catalog No. 50-003-64659