Learn More
Medchemexpress LLC HY-117482 5mg Medchemexpress, BPN-15606 CAS:1914989-49-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1174825MG
Encompass_Preferred
Medchemexpress, HY-117482 5mg BPN-15606 CAS:1914989-49-3 BPN-15606 is a highly potent, orally active γ-secretase modulator (GSM), attenuates the production of Aβ42 and Aβ40 by SHSY5Y neuroblastoma cells with IC50 values of 7 nM and 17nM, respectively. BPN-15606 lowers Aβ42 and Aβ40 levels in the central nervous system of rats and mice. BPN-15606 has acceptable PK/PD properties, including bioavailability, half-life, and clearance. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9828