Learn More
Medchemexpress LLC HY-13537A 5mg Medchemexpress, BPR1J-097 Hydrochloride CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13537A5MG
Encompass_Preferred
Medchemexpress, HY-13537A 5mg BPR1J-097 Hydrochloride CAS: BPR1J-097 Hydrochloride is a novel and potent FLT3 inhibitor with an IC50 of 11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1385