Learn More
Enzo Life Sciences ET A receptor antagonist [BQ-123]. sodium salt (1mg). CAS: 136553-81-6
SDP
Supplier:  Enzo Life Sciences ALX155004P001
Encompass_Preferred
Highly potent and selective ETA antagonist. Formula: C31H42N6O7 . Na. MW: 633.7. Purity: ≥96%. Solubility: Soluble in water. Long Term Storage: -20°C. Use/Stability: Stable for at least 3 years after receipt when stored at -20°C. Stock solutions are stable for up to 3 months when stored at -20°C.
Catalog No. 50-200-7796