Learn More
Enzo Life Sciences ET A receptor antagonist [BQ-610] (1mg). CAS: 141595-53-1
SDP
Supplier:  Enzo Life Sciences ALX155011P001
Encompass_Preferred
30-fold more potent ETA antagonist than BQ-123 in pig aortic smooth muscle membrane assay, same activity as BQ-123 when tested on pig cerebellum membrane. Formula: C36H44N6O6. MW: 656.8. Long Term Storage: -20°C.
Catalog No. 50-200-7802