Learn More
Medchemexpress LLC Br-Boc-C2-azido | 1120364-53-5 | 236.07 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138464100MG
Encompass_Preferred
Br-Boc-C2-azido is an alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing an Azide group, and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Alkyl/ether-based PROTAC linker
Catalog No. 50-003-51017