Learn More
Medchemexpress LLC Br-C10-methyl ester | 6287-90-7 | 97.9% | 10 G
SDP
Supplier:  Medchemexpress LLC HY13064110G
Encompass_Preferred
Br-C10-methyl ester is a PROTAC linker with an alkyl/ether composition, used in the synthesis of various PROTACs. PROTACs are compounds that consist of two distinct ligands connected by a linker, where one ligand targets the VHL protein and the other targets a specific protein.
  • Functions as a PROTAC linker
  • Features an alkyl/ether chemical composition
  • Used in the synthesis of various PROTACs
  • Supports the development of targeted protein degradation
  • Has a molecular weight of 279.21
  • Appears as a solid below 12.5°C and liquid above 12.5°C
  • Colorless to light yellow in appearance
Catalog No. 50-003-40532