Learn More
Medchemexpress LLC Br-C10-methyl ester | 6287-90-7 | 97.9% | 5 G

Supplier: Medchemexpress LLC HY1306415G
Br-C10-methyl ester is a PROTAC linker, characterized by its alkyl/ether composition. It is utilized in the synthesis of PROTACs (MS432), which consist of two distinct ligands connected by a linker: one being the VHL ligand portion and the other for the target protein. It is intended for research use only.
- PROTAC linker with alkyl/ether composition
- Used in the synthesis of PROTACs
- Consists of two distinct ligands connected by a linker
- Appears colorless to light yellow
- Solid below 12.5°C, liquid above 12.5°C
- Soluble in DMSO (100 mg/mL)
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.