Learn More
Medchemexpress LLC Br-C3-methyl ester | 4897-84-1 | 181.03 | 100 G

Supplier: Medchemexpress LLC HYW004701100G
Br-C3-methyl ester is an Alkyl/ether-based PROTAC linker used in the synthesis of PROTAC PD-1/PD-L1 degrader-1. It functions by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- Alkyl/ether-based PROTAC linker
- Purity: 99.1%
- Molecular weight: 181.03
- Formula: C5H9BrO2
- Appearance: Liquid
- Color: Colorless to light yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.