Learn More
Medchemexpress LLC BR-cpd7 | 98.0% | 928.82 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16370510MM/1ML
Encompass_Preferred
BR-cpd7 is a PROTAC degrader designed to target fibroblast growth factor receptor (FGFR1/2). It has a DC50 of 10 nM and works by arresting the cell cycle and inhibiting the proliferation of FGFR1/2 aberrant activated tumor cells.
  • PROTAC degrader for FGFR1/2
  • DC50 of 10 nM
  • Arrests cell cycle
  • Inhibits proliferation of FGFR1/2 aberrant activated tumor cells
Catalog No. 50-003-91704