Learn More
Medchemexpress LLC BR-cpd7 | 98.0% | 928.82 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY163705
Encompass_Preferred
BR-cpd7 is a PROTAC degrader for fibroblast growth factor receptor FGFR1/2 with a DC50 of 10 nM. It arrests cell cycle and inhibits proliferations of FGFR1/2 aberrant activated tumor cells. The compound consists of a ligand for target protein, a linker, and a ligand for E3 ligase.
  • PROTAC degrader for FGFR1/2
  • Has a DC50 of 10 nM
  • Arrests cell cycle
  • Inhibits proliferations of FGFR1/2 aberrant activated tumor cells
Catalog No. 50-003-93278