Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Br-cpd7 | 98.0% | 928.82 | C44H47Cl2N11O8 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY16370510MG

Encompass_Preferred

BR-cpd7 is a PROTAC degrader targeting fibroblast growth factor receptors FGFR1 and FGFR2, exhibiting a DC50 of 10 nM. It arrests the cell cycle and inhibits proliferation in cells with aberrant FGFR1/2 activation. The compound is supplied as a solid powder for research use.

  • Targets FGFR1 and FGFR2 with a DC50 of 10 nM.
  • High purity (98.01%) suitable for research applications.
  • Available in small milligram pack sizes, including 10 MG.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Powder stable at -20°C for up to 3 years; solvent stability varies by condition.

Catalog No. 50-002-89045


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.