Learn More
Medchemexpress LLC Br-PEG3-C2-Boc | 782475-37-0 | 98.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130483
Encompass_Preferred
Br-PEG3-C2-Boc is a PEG-based PROTAC linker designed for the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein via a linker.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-42667