Learn More
Medchemexpress LLC Br-PEG4-C2-Boc | 564476-32-0 | 98.0% | 500 MG
Supplier: Medchemexpress LLC HY130315500MG
Br-PEG4-C2-Boc is a cleavable 4 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Antibody-drug conjugates are comprised of an antibody to which an ADC cytotoxin is attached through an ADC linker.
- Cleavable linker
- PEG-based
- Alkyl/ether structure