Learn More
Medchemexpress LLC Br-PEG4-CH2-Boc | 1807505-29-8 | ≥97.0% | 1 G

Supplier: Medchemexpress LLC HY1305361G
Br-PEG4-CH2-Boc is a PEG- and Alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs consist of two distinct ligands joined by a linker, one targeting an E3 ubiquitin ligase and the other targeting a specific protein. They function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
- PEG- and alkyl/ether-based PROTAC linker
- Utilized in PROTAC synthesis
- Molecular weight: 371.26
- Purity: ≥97.0%
- Appears as a colorless to light yellow liquid
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.