Learn More
Medchemexpress LLC Br-PEG4-OH | 85141-94-2 | 98.2% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1305121G
Encompass_Preferred
Br-PEG4-OH is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins by linking a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker.
  • Utilized in the synthesis of PROTACs.
  • Facilitates the selective degradation of target proteins.
  • Exploits the ubiquitin-proteasome system.
Catalog No. 50-003-40555