Learn More
Medchemexpress LLC Br-PEG6-C2-acid | 309916-91-4 | 417.29 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138362100MG
Encompass_Preferred
Br-PEG6-C2-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase and the other is for the target protein.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-50992