Learn More
Medchemexpress LLC Br-PEG6-C2-acid | 309916-91-4 | 417.29 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138362
Encompass_Preferred
Br-PEG6-C2-acid is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These compounds are utilized in research to selectively degrade target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase and the other targets a protein of interest
  • Utilizes the intracellular ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • For research use only
Catalog No. 50-003-48645