Learn More
Medchemexpress LLC Br-PEG6-C2-acid | 309916-91-4 | 417.29 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13836250MG
Encompass_Preferred
Br-PEG6-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50990