Learn More
Medchemexpress LLC Br-PEG8-OH | 1527515-88-3 | MFCD31656972 | 95.0% | 433.33 g/mol | C16H33BrO8 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1907431G
Encompass_Preferred
Br-PEG8-OH is a brominated polyethylene glycol linker composed of eight ethylene glycol units with a terminal hydroxyl group. It is used as a synthetic building block and linker for chemical synthesis and bioconjugation. Molecular formula: C16H33BrO8. Molecular weight: 433.33 g/mol. CAS number: 1527515-88-3.
  • Brominated polyethylene glycol with eight ethylene glycol units.
  • Terminal hydroxyl group enables further functionalization.
  • Suitable as a linker or building block for chemical synthesis and bioconjugation.
  • Stable under recommended storage conditions for long-term use.
  • Provided with batch certificate/COA for identity confirmation.
Catalog No. 50-002-81927