Learn More
Medchemexpress LLC Bractoppin | 2290527-07-8 | 99.9% | 414.47 g·mol⁻¹ | C25H23FN4O | 25MG
SDP
Supplier:  Medchemexpress LLC HY12602025MG
Encompass_Preferred
Bractoppin is a small-molecule research compound that inhibits phosphopeptide recognition by the BRCA1 tandem BRCT (tBRCT) domain, diminishing BRCA1 recruitment to DNA damage sites and impairing RAD51 assembly. It is provided as a white to off-white solid with high reported purity for biochemical and cellular studies.
  • Potent inhibitor of BRCA1 tBRCT-mediated phosphopeptide recognition.
  • Reduces BRCA1 recruitment to sites of DNA damage.
  • Suppresses RAD51 assembly and damage-induced G2 arrest.
  • High purity suitable for biochemical and cellular research.
  • Supplied as a solid with defined storage recommendations for stability.
Catalog No. 50-002-37973