Learn More
Medchemexpress LLC HY-126020 5mg Medchemexpress, Bractoppin CAS:2290527-07-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1260205MG
Encompass_Preferred
Medchemexpress, HY-126020 5mg Bractoppin CAS:2290527-07-8 Bractoppin is a potent and selective drug-like inhibitor of phosphopeptide recognition by the human BRCA1 tandem(t) BRCT domain (binding IC50: 74 nM). Bractoppin diminishes BRCA1 recruitment to DNA breaks, in turn suppressing damage-induced G2 arrest and assembly of the recombinase, RAD51. Bractoppin preferentially inhibits BRCA1 tBRCT-dependent steps in the DNA damage response. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1090