Learn More
Brady™ BradyJet™ Continuous Vinyl Labels
Brady™ BradyJet™ Continuous Vinyl Labels are ideal for a wide range of identification applications.
Supplier:  Brady™ J20C22502595
 View more versions of this product
Catalog No. 19-129-148
Add to Cart
Description
Description
  • Constructed of indoor-use general purpose vinyl
  • Features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifications
Specifications
Acrylic, Permanent
White
Roll
Single-side Printable
Label
Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking, Lockout Tagout Labeling, GHS and Chemical Labels, Arc Flash Labels
1
1 Label/Bag
-40°C to +60°C
0.135 mm
100 ft.
2.25 in.
1.88 in. I.D.
None
Matte
Vinyl
Ink Jet
Non-sterile
BradyJet™ J2000 Printer
-40°C
-40°F to +140°F
0.0053 in.
100 ft. x 2.25 in.
30.48 m
57.15 mm
J20C-2250-2595
Product Suggestions
Product Suggestions