Learn More
Brady™ BradyJet™ J2000 Repositionable Inkjet Vinyl Labels
The special aggressive adhesive on this Repositionable Inkjet Material stays in place when you need it to, but removes cleanly without leaving adhesive residue making it ideal for 5S and inventory applications.
Supplier:  Brady™ J20C22502581
 View more versions of this product
Catalog No. 19-129-503
Add to Cart
Description
Description
  • Durable and flexible material making it ideal for areas where identification is frequently changed
  • Service temperature of -40 to 140°F (-40 to 60°C)
Specifications
Specifications
Acrylic, Removable
White
Continuous
Single-side Printable
Label
Temporary Labeling
1
1 Label/Bag
-40°C to +60°C
0.254 mm
100 ft.
2.25 in.
J20C-2250-2581
None
Matte
Vinyl
Ink Jet
Non-sterile
BradyJet™ J2000
-40°C
-40°F to +140°F
0.01 in.
100 ft. x 2.25 in.
30.48 m
57.15 mm
Product Suggestions
Product Suggestions