Learn More
Bachem Bradykinin Acetate
SDP
Supplier:  Bachem 4002961.0100
Encompass_Preferred
Bradykinin Acetate, H-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH, 6846-03-3, C50H73N15O11, Mr 1060.22, 100mg. Kallidin I, BK, Kallidin-9, BRS-640. The pharmacological properties of bradykinin include smooth muscle contraction, vasodilation and hypotension. Bradykinin increases capillary permeability together with edema formation and induction of pain. Bradykinin induces bronchoconstriction in asthmatic subjects and symptoms of rhinitis in nasal provocation. CAS Number (net): 58-82-2.
Catalog No. NC9481566