Learn More
Medchemexpress LLC Bradykinin (2-9) | 16875-11-9 | 97.15% | 904.02 | 1 MG

Supplier: Medchemexpress LLC HYP14901MG
Bradykinin (2-9) is an amino-truncated Bradykinin peptide and a metabolite of Bradykinin, cleaved by Aminopeptidase P.
- Molecular weight: 904.02
- Formula: C44H61N11O10
- Sequence: Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg
- Appearance: Solid, white to off-white
- Solubility in DMSO: 100 mg/mL (110.62 mM)
- Below limits of detection in rat kidney and blood in a tissue-based system
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.