Learn More
Cayman Chemical BradykInIn Fragment1-5tr 5mg
SDP
Supplier:  Cayman Chemical 353025 MG
Encompass_Preferred
A metabolite of bradykinin; increases NO production in neonatal rat cardiomyocytes at 100 nM; inhibits α-thrombin-induced platelet aggregation of washed isolated platelets (IC50 = 0.5 mM); prevents decreases in mean arterial blood pressure and heart rate and increases survival in a rat model of LPS-induced septicemia
Catalog No. 50-290-9550